Skip to Content
Merck
All Photos(1)

Key Documents

465038

Sigma-Aldrich

Beryllium nitrate solution

35 wt. % in H2O, 99.99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Be(NO3)2
CAS Number:
Molecular Weight:
133.02
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
99.99%

Assay

99.99%

concentration

35 wt. % in H2O

density

1.2 g/mL at 25 °C

SMILES string

[Be++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Be.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

RFVVBBUVWAIIBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B Inhalation - Eye Irrit. 2 - Ox. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satendra Kumar Nirala et al.
Indian journal of experimental biology, 47(12), 955-963 (2010-03-25)
To evaluate therapeutic efficacy of chelating agents tiron (Sodium-4,5-dihydroxy-1,3-benzene disulphonate) and CaNa3DTPA (Calcium trisodium diethylene triamine pentaacetic acid) in presence of alpha-tocopherol against beryllium induced toxicity, adult female albino rats were exposed to beryllium nitrate for 28 days followed by
R Mathur et al.
Indian journal of experimental biology, 42(6), 570-574 (2004-07-21)
The therapeutic efficacy of chelating agents CaNa3DTPA (calcium trisodium diethylene triamine penta acetic acid) and Tiron (sodium-4,5-dihydroxy-1,3-benzene disulphonate) with and without antioxidant, alpha-Tocopherol was evaluated in the treatment of beryllium-induced toxicity in female albino rats. The animals were exposed to
Effect of beryllium nitrate on early and late pregnancy in rats.
R Mathur et al.
Bulletin of environmental contamination and toxicology, 38(1), 73-77 (1987-01-01)
Satendra Kumar Nirala et al.
Journal of applied toxicology : JAT, 28(1), 44-54 (2007-04-13)
The therapeutic potential of the chelator tiferron (sodium-4,5-dihydroxy-1,3-benzene disulphonate; 300 mg kg(-1), i.p.) and adjuvants, i.e. alpha-tocopherol (25 mg kg(-1), p.o.), propolis (a honey-bee hive product; 200 mg kg(-1), p.o.) and piperine (10 mg kg(-1), p.o.) were evaluated individually and
A O Prakash et al.
Journal of ethnopharmacology, 18(2), 143-146 (1986-11-01)
The feeding of powdered leaves of Gymnema sylvestre in the diet of rats for 10 days prior and 15 days after i.v. beryllium nitrate significantly protected the animals from the full fall of blood glucose seen in rats receiving beryllium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service