Skip to Content
Merck

Skip To

463531

Tris[2-(methylamino)ethyl]amine

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
$221.00
25 g
Check Cart for Availability
$806.00

About This Item

Linear Formula:
(H3CNHCH2CH2)3N
CAS Number:
Molecular Weight:
188.31
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
97%

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

bp

77-78 °C/0.1 mmHg (lit.)

density

0.896 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNCCN(CCNC)CCNC

InChI

1S/C9H24N4/c1-10-4-7-13(8-5-11-2)9-6-12-3/h10-12H,4-9H2,1-3H3

InChI key

GMLHUKQBMDKQBD-UHFFFAOYSA-N

General description

Tris[2-(methylamino)ethyl]amine is a polyamine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
645095549835102245
assay

97%

assay

90%

assay

96%

assay

97%

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

density

0.896 g/mL at 25 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

density

-

density

0.984 g/mL at 25 °C (lit.)

bp

77-78 °C/0.1 mmHg (lit.)

bp

120 °C/0.5 mmHg (lit.)

bp

-

bp

191 °C (lit.)

functional group

amine

functional group

amine

functional group

amine

functional group

amine


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

228.0 °F - closed cup

flash_point_c

108.9 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters