Skip to Content
Merck
All Photos(1)

Key Documents

462845

Sigma-Aldrich

2-Butyl-1-indanone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O
CAS Number:
Molecular Weight:
188.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.531 (lit.)

bp

98 °C/0.5 mmHg (lit.)

density

1.002 g/mL at 25 °C (lit.)

SMILES string

CCCCC1Cc2ccccc2C1=O

InChI

1S/C13H16O/c1-2-3-6-11-9-10-7-4-5-8-12(10)13(11)14/h4-5,7-8,11H,2-3,6,9H2,1H3

InChI key

AWYXNIWXOXKRDQ-UHFFFAOYSA-N

General description

2-Butyl-1-indanone is an indanone derivative. It undergoes transfer hydrogenation reaction with tributylamine in the presence of Pd/C catalyst.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and substituent effects on retention and chiral resolution of ketones and alcohols on microcrystalline cellulose triacetate plates.
Lepri L, et al.
Chromatographia, 71(7-8), 685-694 (2010)
Transfer hydrogenation of ketones to alkanes
using tributylamine under heterogeneous Pd/C
catalysis
Kim TG, et al.
Applied Organometallic Chemistry, 28(10), 797-799 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service