Skip to Content
Merck
All Photos(1)

Key Documents

459313

Sigma-Aldrich

Boc-Cys(Trt)-OH

97%, for peptide synthesis

Synonym(s):

N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, Nα-Boc-S-trityl-L-cysteine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$94.40
25 G
$396.00

$94.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$94.40
25 G
$396.00

About This Item

Linear Formula:
(C6H5)3CSCH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
463.59
Beilstein:
2028965
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$94.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Boc-Cys(Trt)-OH, 97%

Assay

97%

optical activity

[α]20/D +27.5°, c = 1 in ethanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

143-146 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CSC(c1ccccc1)(c2ccccc2)c3ccccc3)C(O)=O

InChI

1S/C27H29NO4S/c1-26(2,3)32-25(31)28-23(24(29)30)19-33-27(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18,23H,19H2,1-3H3,(H,28,31)(H,29,30)/t23-/m0/s1

InChI key

JDTOWOURWBDELG-QHCPKHFHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sunithi Gunasekera et al.
International journal of peptide research and therapeutics, 19(1), 43-54 (2013-03-19)
The development of synthetic methodologies for cyclic peptides is driven by the discovery of cyclic peptide drug scaffolds such as the plant-derived cyclotides, sunflower trypsin inhibitor 1 (SFTI-1) and the development of cyclized conotoxins. Currently, the native chemical ligation reaction
Yang Zhang et al.
ACS applied materials & interfaces, 11(31), 27529-27535 (2019-07-11)
Characterizing over-expressed enzymes or biomarkers in living cells is critical for the molecular understanding of disease pathology and consequently for designing precision medicines. Herein, a "switch-on" probe is designed to selectively detect γ-glutamyl transpeptidase (GGT) in living cells via a
Naohisa Ogo et al.
Bioorganic & medicinal chemistry letters, 17(14), 3921-3924 (2007-05-26)
Inhibition of Eg5 represents a novel approach for the treatment of cancer. Here, we report the synthesis and structure-activity relationship of S-trityl-L-cysteine (STLC) derivatives as Eg5 inhibitors. Some of these derivatives such as 4f demonstrated enhanced inhibitory activity against Eg5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service