Skip to Content
Merck
All Photos(1)

Key Documents

459291

Sigma-Aldrich

Fmoc-Nle-OH

97%

Synonym(s):

(S)-2-(Fmoc-amino)caproic acid, (S)-2-(Fmoc-amino)hexanoic acid, N-(9-Fluorenylmethoxycarbonyl)-L-norleucine, Fmoc-L-norleucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Beilstein:
5305164
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −18.5°, c = 1 in DMF

mp

141-144 °C (lit.)

SMILES string

CCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4/c1-2-3-12-19(20(23)24)22-21(25)26-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,2-3,12-13H2,1H3,(H,22,25)(H,23,24)/t19-/m0/s1

InChI key

VCFCFPNRQDANPN-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service