Skip to Content
Merck
All Photos(1)

Key Documents

458627

Sigma-Aldrich

Poly[bis(2-chloroethyl) ether-alt-1,3-bis[3-(dimethylamino)propyl]urea] quaternized, solution

62 wt. % in H2O

Synonym(s):

Polyquaternium-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$87.10

$87.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$87.10

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$87.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

concentration

62 wt. % in H2O

refractive index

n20/D 1.468

pH

7.5-8.5 (10 wt. % in solution)

bp

105 °C

density

1.13 g/mL at 25 °C

Application

Adhesion promoter toward polar surfaces. Conditioning agent in personal care products.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Questions

1–2 of 2 Questions  
  1. has polyquad-2 an antimicrobial activity?

    1 answer
    1. This product is not tested for antimicrobial activity. There are very few publications that reference this compound as a preservative or biocidal agent. See the link below for an article that may be of interest.
      https://journals.sagepub.com/doi/abs/10.1177/0883911515627473

      Helpful?

  2. what is the molecular weight of this polymer? (Poly[bis(2-chloroethyl) ether-alt-1,3-bis[3-(dimethylamino)propyl]urea] quaternized, solution)

    1 answer
    1. The average molecular weight is >10,000.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service