Skip to Content
Merck
All Photos(1)

Key Documents

458554

Sigma-Aldrich

(3S-cis)-(+)-2,3-Dihydro-3-isopropyl-7a-methylpyrrolo[2,1-b]oxazol-5(7aH)-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +41.0°, c = 1 in chloroform

mp

46-49 °C (lit.)

SMILES string

CC(C)[C@H]1CO[C@]2(C)C=CC(=O)N12

InChI

1S/C10H15NO2/c1-7(2)8-6-13-10(3)5-4-9(12)11(8)10/h4-5,7-8H,6H2,1-3H3/t8-,10-/m1/s1

InChI key

JOXQDNAGAGXGOK-PSASIEDQSA-N

Application

High levels of asymmetric induction have been obtained with this chiral bicyclic lactam in cycloadditions, alkylations, conjugate additions, annulations, and dihydroxylations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romo, D. Meyers, A.I.
Tetrahedron, 47, 9503-9503 (1991)
Andres, C.J. et al.
Tetrahedron Letters, 60, 3189-3189 (1995)
Brengel, G.P. Meyers, A.I.
The Journal of Organic Chemistry, 61, 3230-3230 (1996)
Munchhoff, M.J. Meters, A.I.
The Journal of Organic Chemistry, 60, 7086-7086 (1995)
Sandham, D.A. Meyers, A.I.
Journal of the Chemical Society. Chemical Communications, 2511-2511 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service