Skip to Content
Merck
All Photos(1)

Key Documents

457779

Sigma-Aldrich

DL-Dithiothreitol

99%

Synonym(s):

threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719757
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

99%

mp

41-44 °C (lit.)

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueqian Liu et al.
The Journal of biological chemistry, 288(10), 7230-7240 (2013-01-25)
In Pichia pastoris, the peroxisomal targeting signal 2 (PTS2)-dependent peroxisomal matrix protein import pathway requires the receptor, Pex7, and its co-receptor Pex20. A conserved lysine (Lys(19)) near the N terminus of Pex20 is required for its polyubiquitination and proteasomal degradation
Jin-Gu Lee et al.
Nature communications, 4, 1568-1568 (2013-03-07)
In eukaryotes, deubiquitinases (DUBs) remove ubiquitin conjugates from diverse substrates, altering their stabilities, localizations or activities. Here we show that many DUBs of the USP and UCH subfamilies can be reversibly inactivated upon oxidation by reactive oxygen species in vitro
Mojtaba Abdul Roda et al.
PloS one, 9(5), e97594-e97594 (2014-05-20)
A novel neutrophil chemoattractant derived from collagen, proline-glycine-proline (PGP), has been recently characterized in chronic obstructive pulmonary disease (COPD). This peptide is derived via the proteolytic activity of matrix metalloproteases (MMP's)-8/9 and PE, enzymes produced by neutrophils and present in
Je-Hung Kuo et al.
The Journal of biological chemistry, 288(19), 13522-13533 (2013-03-29)
CXCL4L1 is a highly potent anti-angiogenic and anti-tumor chemokine, and its structural information is unknown. CXCL4L1 x-ray structure is determined, and it reveals a previously unrecognized chemokine structure adopting a novel C-terminal helix conformation. The alternative helix conformation enhances the
František Štork et al.
Environmental science and pollution research international, 20(8), 5502-5511 (2013-02-26)
Effect of nitric oxide donor (sodium nitroprusside, SNP, 500 μM) or hydrogen peroxide scavenger (dithiothreitol, DTT, 500 μM) on cadmium (Cd) or copper (Cu) uptake (150 μM solutions) and toxicity using Scenedesmus quadricauda was studied. Combined treatments (Cd or Cu

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service