Skip to Content
Merck
All Photos(1)

Key Documents

457752

Sigma-Aldrich

2-Methyl-4H-3,1-benzoxazin-4-one

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$81.40

$81.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$81.40

About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$81.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

79-82 °C (lit.)

functional group

ester

SMILES string

CC1=Nc2ccccc2C(=O)O1

InChI

1S/C9H7NO2/c1-6-10-8-5-3-2-4-7(8)9(11)12-6/h2-5H,1H3

InChI key

WMQSKECCMQRJRX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Simple Synthesis of 2-Substituted-4 H-3, 1-benzoxazin-4-ones by Palladium-Catalyzed Cyclocarbonylation of o-Iodoanilines with Acid Chlorides.
Chitchamai L and Alper H.
Organic Letters, 1(10), 1619-1622 (1999)
B Siedle et al.
Bioorganic & medicinal chemistry, 10(9), 2855-2861 (2002-07-12)
Human neutrophil elastase (HNE) is a serine protease that has been implicated in the abnormal turnover of connective tissue proteins and has been described as an important pathogenic factor in several inflammatory diseases such as rheumatoid arthritis or cystic fibrosis.
Reactivity of 4H-3,1-benzoxazin-4-ones towards nitrogen and carbon nucleophilic reagents: applications to the synthesis of new heterocycles.
Madkour HMF.
ARKIVOC (Gainesville, FL, United States), 1, 36-54 (2004)
L Hedstrom et al.
Biochemistry, 23(8), 1753-1759 (1984-04-10)
The benzoxazinones 2-ethoxy-4H-3,1- benzoxazin -4-one (1a) and 2-(trifluoromethyl)-4H-3,1- benzoxazin -4-one (1d) inactivate chymotrypsin. The inactivation is stoichiometric and proceeds with rate constants of 7 X 10(5) M-1 min-1 and greater than 4 X 10(6) M-1 min-1, respectively. The inactivated enzyme

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service