Skip to Content
Merck
All Photos(1)

Key Documents

457345

Sigma-Aldrich

2′,4′-Dichloro-5′-fluoroacetophenone

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(F)COCH3
CAS Number:
Molecular Weight:
207.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

refractive index

n20/D 1.546 (lit.)

bp

167 °C (lit.)

density

1.425 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cc(F)c(Cl)cc1Cl

InChI

1S/C8H5Cl2FO/c1-4(12)5-2-8(11)7(10)3-6(5)9/h2-3H,1H3

InChI key

FAKJFAMIABOKBW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B S Holla et al.
Farmaco (Societa chimica italiana : 1989), 56(8), 565-570 (2001-10-17)
A series of 7-arylidene-6-(2,4-dichloro-5-fluorophenyl)-3-substituted-1,2,4-triazolo[3,4-b]-1,3,4-thiadiazines (3) were prepared by the condensation of 4-amino-5-mercapto-3-substituted-1,2,4-triazoles (1) and 3-aryl-1-(2,4-dichloro-5-fluorophenyl)-2-bromo-2-propen-1-one (2). An alternative route for the synthesis of the title compound 3 has been described. The newly synthesised compounds were characterised on the basis of
Journal of Heterocyclic Chemistry, 25, 927-927 (1988)
D T Chu et al.
Journal of medicinal chemistry, 28(11), 1558-1564 (1985-11-01)
A series of novel arylfluoroquinolones has been prepared. These derivatives are characterized by having a fluorine atom at the 6-position, substituted amino groups at the 7-position, and substituted phenyl groups at the 1-position. Structure-activity relationship (SAR) studies indicate that the
Eco-friendly synthesis of fluorine-containing pyrazoline derivatives over potassium carbonate.
Patel VM and Desai KR.
ARKIVOC (Gainesville, FL, United States), 1, 123-129 (2004)
Zhongguo Yiyao Gongye Zazhi, 25, 296-296 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service