Skip to Content
Merck
All Photos(1)

Documents

457027

Sigma-Aldrich

3,4-Difluorohydrocinnamic acid

97%

Synonym(s):

3-(3,4-Difluorophenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2CH2CO2H
CAS Number:
Molecular Weight:
186.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

49-53 °C (lit.)

SMILES string

OC(=O)CCc1ccc(F)c(F)c1

InChI

1S/C9H8F2O2/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1,3,5H,2,4H2,(H,12,13)

InChI key

UOZIYCHJMUNLIG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorna Piazzi et al.
Bioorganic & medicinal chemistry letters, 18(1), 423-426 (2007-11-14)
The complex etiology of Alzheimer's disease (AD) prompts scientists to develop multifunctional compounds to combat causes and symptoms of such neurodegeneration. To this aim we designed, synthesized, and tested a series of compounds by introducing halophenylalkylamidic functions on the scaffold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service