Skip to Content
Merck
All Photos(1)

Key Documents

456861

Sigma-Aldrich

4-Chlorophenoxyacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OCH2CN
CAS Number:
Molecular Weight:
167.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

45-49 °C (lit.)

functional group

chloro
nitrile

SMILES string

Clc1ccc(OCC#N)cc1

InChI

1S/C8H6ClNO/c9-7-1-3-8(4-2-7)11-6-5-10/h1-4H,6H2

InChI key

YUGDKEWUYZXXRU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel and selective 5-HT2C/2B receptor antagonists as potential anxiolytic agents: synthesis, quantitative structure-activity relationships, and molecular modeling of substituted 1-(3-pyridylcarbamoyl) indolines.
Bromidge SM, et al.
Journal of Medicinal Chemistry, 41(10), 1598-1612 (1998)
Synthesis of C3-Substituted 4-Azaindoles: An Easy Access to 4-Azamelatonin and Protected 4-Azatryptophan.
Jeanty M, et al.
The Journal of Organic Chemistry, 73(!8), 7390-7393 (2008)
Vlachou M, et al.
Heterocycles, 129-132 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service