Skip to Content
Merck
All Photos(1)

Documents

456136

Sigma-Aldrich

(R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClO2
CAS Number:
Molecular Weight:
150.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]24/D +42.1°, neat

refractive index

n20/D 1.434 (lit.)

bp

63 °C/37 mmHg (lit.)

density

1.103 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OC[C@H](CCl)O1

InChI

1S/C6H11ClO2/c1-6(2)8-4-5(3-7)9-6/h5H,3-4H2,1-2H3/t5-/m0/s1

InChI key

BNPOTXLWPZOESZ-YFKPBYRVSA-N

Application

Utilized in the preparation of both enantiomers of the orally active antifungal azolic agent ketoconazole.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin R Campos et al.
Journal of the American Chemical Society, 128(11), 3538-3539 (2006-03-16)
This communication discloses the first instance of the enantioselective Pd-catalyzed alpha-arylation of N-Boc-pyrrolidine. The methodology relies on Beak's sparteine-mediated, enantioselective deprotonation of N-Boc-pyrrolidine to form the 2-pyrrolidinolithium specices in high enantiomeric ratio (er). Transmetalation of this intermediate with zinc chloride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service