Skip to Content
Merck
All Photos(1)

Key Documents

455040

Sigma-Aldrich

Dimethyl (R)-(+)-malate

98%

Synonym(s):

Dimethyl (R)-2-hydroxysuccinate, Dimethyl D-malate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$345.00

$345.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$345.00

About This Item

Linear Formula:
CH3O2CCH2CH(OH)CO2CH3
CAS Number:
Molecular Weight:
162.14
Beilstein:
1908705
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$345.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]20/D +6.6°, neat

refractive index

n20/D 1.44 (lit.)

bp

104-108 °C/1 mmHg (lit.)

density

1.232 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

storage temp.

2-8°C

SMILES string

COC(=O)C[C@@H](O)C(=O)OC

InChI

1S/C6H10O5/c1-10-5(8)3-4(7)6(9)11-2/h4,7H,3H2,1-2H3/t4-/m1/s1

InChI key

YSEKNCXYRGKTBJ-SCSAIBSYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service