Skip to Content
Merck
All Photos(1)

Key Documents

452114

Sigma-Aldrich

Ethyl bromofluoroacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH(F)CO2C2H5
CAS Number:
Molecular Weight:
184.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.424 (lit.)

bp

154 °C (lit.)

density

1.565 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C(F)Br

InChI

1S/C4H6BrFO2/c1-2-8-4(7)3(5)6/h3H,2H2,1H3

InChI key

ULNDTPIRBQGESN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Fluorine Chemistry, 77, 45-45 (1996)
R Ocampo et al.
The Journal of organic chemistry, 67(1), 72-78 (2002-01-05)
The presence of catalytic amounts of CeCl(3) improves yields and simplifies procedure in the Reformatsky reactions of ethyl bromofluoroacetate with aldehydes and ketones to generate diastereomeric mixtures of alpha-fluoro-beta-hydroxy esters, some of which can be separated by crystallization or column
Thibault Ferrary et al.
Organic letters, 15(21), 5598-5601 (2013-10-22)
The synthesis of highly functionalized monofluorinated cyclopropanes based on a Michael Initiated Ring Closure (MIRC) reaction has been developed. The addition of quaternary ammonium salts derived from ethyl bromofluoroacetate on a panel of electron deficient alkenes followed by cyclization gave
Xiaoyang Sun et al.
Organic letters, 16(11), 2938-2941 (2014-05-23)
A practical and unified strategy has been described for the preparation of mono- and difluoromethylated phenanthridine derivatives using a visible-light-promoted alkylation and decarboxylation sequence from biphenyl isocyanides with ethyl bromofluoroacetate (EBFA) or ethyl bromodifluoroacetate (EBDFA). These reactions could be carried
Synthesis of (E)-and (Z)-fluoro-olefin analogues of potent dipeptidyl peptidase IV inhibitors.
Van der Veken P, et al.
Tetrahedron Letters, 44(33), 6231-6234 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service