Skip to Content
Merck
All Photos(2)

Documents

449822

Sigma-Aldrich

Lanthanum(III) bromide

AnhydroBeads, −10 mesh, ≥99.99% trace metals basis

Synonym(s):

Lanthanum bromide, Lanthanum tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaBr3
CAS Number:
Molecular Weight:
378.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

≥99.99% trace metals basis

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤100.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

783 °C (lit.)

density

5.06 g/mL at 25 °C (lit.)

SMILES string

Br[La](Br)Br

InChI

1S/3BrH.La/h3*1H;/q;;;+3/p-3

InChI key

XKUYOJZZLGFZTC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Applied in solid state synthesis of CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units.

Features and Benefits

Applied in solid state synthesis of CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lulei, M.
Inorganic Chemistry, 37, 777-777 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service