Skip to Content
Merck
All Photos(1)

Key Documents

449806

Sigma-Aldrich

Europium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Europium fluoride, Europium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
EuF3
CAS Number:
Molecular Weight:
208.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: europium

impurities

≤150.0 ppm Trace Rare Earth Analysis

bp

2280 °C (lit.)

SMILES string

F[Eu](F)F

InChI

1S/Eu.3FH/h;3*1H/q+3;;;/p-3

InChI key

HPNURIVGONRLQI-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of novel mixed-metal fluorides and in studies on fluoride glasses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeitschrift fur Anorganische und Allgemeine Chemie, 622, 1601-1601 (1996)
Ferroelectr., Lett. Sect., 20, 59-59 (1995)
Journal of Luminescence, 66, 290-290 (1995)
Journal of Non-Crystalline Solids, 197, 8-8 (1996)
R Peslin et al.
Journal of applied physiology (Bethesda, Md. : 1985), 80(6), 2198-2203 (1996-06-01)
Specific airway resistance (sRaw) measured by body plethysmography without conditioning of the inspired air to BTPS exhibits a strong frequency dependence related to the fact that the warming and wetting of the gas in the airways is not instantaneous (R.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service