Skip to Content
Merck
All Photos(1)

Key Documents

448524

Sigma-Aldrich

Trichloronitromethane

98%

Synonym(s):

Chloropicrin, Nitrotrichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CNO2
CAS Number:
Molecular Weight:
164.38
Beilstein:
1756135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

98%

refractive index

n20/D 1.461 (lit.)

bp

112 °C (lit.)

density

1.657 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)C(Cl)(Cl)Cl

InChI

1S/CCl3NO2/c2-1(3,4)5(6)7

InChI key

LFHISGNCFUNFFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service