447641
Germanium(IV) isopropoxide
97%
Synonym(s):
Germanium tetraisopropoxide, Isopropyl germanate(IV), Tetraisopropoxygermane, Tetraisopropoxygermanium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Ge(OCH(CH3)2)4
CAS Number:
Molecular Weight:
308.99
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
reaction suitability
core: germanium
reagent type: catalyst
bp
167 °C (lit.)
density
1.033 g/mL at 25 °C (lit.)
SMILES string
CC(C)O[Ge](OC(C)C)(OC(C)C)OC(C)C
InChI
1S/C12H28GeO4/c1-9(2)14-13(15-10(3)4,16-11(5)6)17-12(7)8/h9-12H,1-8H3
InChI key
PKLMYPSYVKAPOX-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
140.0 °F - closed cup
Flash Point(C)
60 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service