Skip to Content
Merck
All Photos(1)

Key Documents

447226

Sigma-Aldrich

Lanthanum(III) hydroxide

99.9% trace metals basis

Synonym(s):

Lanthanum hydroxide, Lanthanum trihydroxide, Lanthanum(III) hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(OH)3
CAS Number:
Molecular Weight:
189.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
powder

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤2000.0 ppm Trace Rare Earth Analysis

SMILES string

O[La](O)O

InChI

1S/La.3H2O/h;3*1H2/q+3;;;/p-3

InChI key

YXEUGTSPQFTXTR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Domínguez et al.
Journal of pineal research, 8(4), 367-374 (1990-01-01)
Lanthanum hydroxide together with a fixative solution was injected into the third ventricle of the viscacha. The tracer penetrated freely into the pineal gland demonstrating an extensive network of intercommunicating intercellular spaces. Numerous gap junctions were observed indicating that they
I V Gmoshinskiĭ et al.
Biulleten' eksperimental'noi biologii i meditsiny, 103(2), 161-163 (1987-02-01)
Adult rats with experimental vitamin A deficiency and control animals were intraperitoneally injected with chicken ovalbumin (OA) solution and the entrance of native OA into the blood was assessed 3 hours later by competitive radioimmunoassay. The OA amounts circulating in
Choon-Ki Na et al.
Journal of hazardous materials, 183(1-3), 512-520 (2010-08-13)
This research was undertaken to evaluate the feasibility of lanthanum hydroxide for fluoride removal from aqueous solutions. A batch sorption experiments were conducted to study the influence of various factors such as pH, presence of competing anions, contact time, initial
Felipe Medrano et al.
Chemical communications (Cambridge, England), (15)(15), 1968-1970 (2003-08-23)
Glycine and N,N-dimethylglycine stabilize La(III) hydroxide complexes of the type La2L2(OH)4 which possess phosphodiesterolytic activity close to that observed with most active tetravalent cations like Ce(IV).
D Bommannan et al.
Pharmaceutical research, 9(8), 1043-1047 (1992-08-01)
We have shown previously that high-frequency ultrasound (sonophoresis) can significantly enhance the transdermal delivery of a topically applied drug in vivo and that the augmentation of transport was caused by the action of the ultrasound on the skin. However, these

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service