Skip to Content
Merck
All Photos(1)

Key Documents

446572

Sigma-Aldrich

2,3,6-Trifluorophenylacetic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.70

$89.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$89.70

About This Item

Linear Formula:
F3C6H2CH2CO2H
CAS Number:
Molecular Weight:
190.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$89.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

111-114 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)Cc1c(F)ccc(F)c1F

InChI

1S/C8H5F3O2/c9-5-1-2-6(10)8(11)4(5)3-7(12)13/h1-2H,3H2,(H,12,13)

InChI key

QRAZASHLGLHKEB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Primary ion production from various gases in spontaneous desorption.
Park MA and Schweikert EA.
International Journal of Mass Spectrometry and Ion Processes, 128(1), 107-113 (1993)
Surfactant-free emulsion electrosynthesis via power ultrasound: electrocatalytic formation of carbon-carbon bonds.
Davies TJ, et al.
Green Chemistry, 4(6), 570-577 (2002)
New C~ 2-Fragment for Ruthenium-Catalyzed Synthesis of Indoles.
Cho CS, et al.
Bull. Korean Chem. Soc., 28(5), 832-832 (2007)
Cyclodextrins as chiral stationary phases in capillary gas chromatography. Part IV: Heptakis (2, 3, 6-tri-O-pentyl)-?-cyclodextrin.
Konig WA, et al.
Journal of High Resolution Chromatography, 12(1), 35-39 (1989)
Synthesis of 2, 2-Dialkylcyclopropane Nitriles1.
Nelson ER, et al.
Journal of the American Chemical Society, 79(13), 3467-3469 (1957)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service