Skip to Content
Merck
All Photos(1)

Key Documents

445355

Sigma-Aldrich

(S)-(+)-1-Aminoindan

97%

Synonym(s):

(S)-(+)-1-Indanamine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$146.00
1 G
$234.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$146.00
1 G
$234.00

About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
Beilstein:
2206708
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

97%

form

liquid

optical activity

[α]20/D +16.5°, c = 1.5 in methanol

refractive index

n20/D 1.562 (lit.)

bp

96-97 °C/8 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

N[C@H]1CCc2ccccc12

InChI

1S/C9H11N/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4,9H,5-6,10H2/t9-/m0/s1

InChI key

XJEVHMGJSYVQBQ-VIFPVBQESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service