Skip to Content
Merck
All Photos(1)

Key Documents

445029

Sigma-Aldrich

cis-5-Dodecenoic acid

99%

Synonym(s):

(5Z)-5-Dodecenoic acid, (Z)-Dodec-5-enoic acid, cis-5-Dodecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CH(CH2)3CO2H
CAS Number:
Molecular Weight:
198.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.454 (lit.)

bp

135 °C/0.4 mmHg (lit.)

density

0.906 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

[H]\C(CCCCCC)=C(/[H])CCCC(O)=O

InChI

1S/C12H22O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h7-8H,2-6,9-11H2,1H3,(H,13,14)/b8-7-

InChI key

IJBFSOLHRKELLR-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidant and cyclooxygenase activities of fatty acids found in food.
Henry GE, et al.
Journal of Agricultural and Food Chemistry, 50(8), 2231-2234 (2002)
Decenoic, dodecenoic, and tetradecenoic acids in the Lactobacteriaceae.
W M O'Leary
Biochemistry, 4(8), 1621-1627 (1965-08-01)
A revised pathway for the oxidation of oleic acid and odd-numbered double bond unsaturated fatty acids.
K Y Tserng et al.
Progress in clinical and biological research, 375, 263-270 (1992-01-01)
Youjun Feng et al.
The Journal of biological chemistry, 284(43), 29526-29535 (2009-08-15)
Although the unsaturated fatty acid (UFA) synthetic pathway of Escherichia coli is the prototype of such pathways, several unresolved issues have accumulated over the years. The key players are the fabA and fabB genes. Earlier studies of fabA transcription showed
K Y Tserng et al.
The Biochemical journal, 308 ( Pt 1), 39-44 (1995-05-15)
The metabolism of cis-5 unsaturated fatty acids was studied in intact rat liver mitochondria to assess the operation of a reduction pathway. By using direct quantification of metabolites with a capillary-column gas chromatography, 3-hydroxydodecanoate was identified among other metabolites when

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service