Skip to Content
Merck
All Photos(1)

Key Documents

444839

Sigma-Aldrich

4(5)-(Iodoacetamido)fluorescein

Dye content 90 %

Synonym(s):

IAF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14INO6
CAS Number:
Molecular Weight:
515.25
Beilstein:
6543244
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 90%

mp

265-267 °C (dec.) (lit.)

λmax

266 nm in DMF

SMILES string

OC(=O)c1ccc(NC(=O)CI)cc1C2=C3C=CC(=O)C=C3Oc4cc(O)ccc24

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Vito Moretti
Frontiers in neurology, 6, 152-152 (2015-07-29)
Recent studies demonstrate that the alpha3/alpha2 power ratio correlates with cortical atrophy, regional hypoperfusion, and memory impairment in subjects with mild cognitive impairment (MCI). Evidences were reviewed in subjects with MCI, who underwent EEG recording, magnetic resonance imaging (MRI) scans
Claudio Babiloni et al.
NeuroImage. Clinical, 12, 144-156 (2016-07-14)
Here we tested the effect of combined antiretroviral therapy (cART) on deviant electroencephalographic (EEG) source activity in treatment-naïve HIV individuals. Resting state eyes-closed EEG data were recorded before and after 5 months of cART in 48 male HIV subjects, who were
Roberta Lizio et al.
Journal of Alzheimer's disease : JAD, 49(1), 159-177 (2015-10-08)
Here we presented a single electroencephalographic (EEG) marker for a neurophysiological assessment of Alzheimer's disease (AD) patients already diagnosed by current guidelines. The ability of the EEG marker to classify 127 AD individuals and 121 matched cognitively intact normal elderly
Anja Pahor et al.
The Spanish journal of psychology, 19, E88-E88 (2016-12-06)
A brief overview of structural and functional brain characteristics related to g is presented in the light of major neurobiological theories of intelligence: Neural Efficiency, P-FIT and Multiple-Demand system. These theories provide a framework to discuss the main objective of
Claudio Babiloni et al.
Clinical neurophysiology : official journal of the International Federation of Clinical Neurophysiology, 127(11), 3455-3469 (2016-10-27)
Cortical sources of electroencephalographic (EEG) rhythms were investigated in two sub-populations of naïve HIV subjects, grouped based on clinical criteria to receive different combination anti-retroviral therapies (cARTs). These EEG sources were hypothesized to reflect beneficial effects of both regimes. Eyes-closed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service