Skip to Content
Merck
All Photos(1)

Key Documents

444391

Sigma-Aldrich

4-(1H-Imidazol-1-yl)aniline

98%

Synonym(s):

1-(4-Aminophenyl)-1H-imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$99.40
5 G
$463.00

$99.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$99.40
5 G
$463.00

About This Item

Empirical Formula (Hill Notation):
C9H9N3
CAS Number:
Molecular Weight:
159.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$99.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

143-147 °C (lit.)

SMILES string

Nc1ccc(cc1)-n2ccnc2

InChI

1S/C9H9N3/c10-8-1-3-9(4-2-8)12-6-5-11-7-12/h1-7H,10H2

InChI key

LVOASPZGXNAHJI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing-Hong Shi et al.
Journal of chromatography. A, 1216(33), 6081-6087 (2009-07-18)
4-(1H-imidazol-1-yl) aniline (AN) was immobilized on Sepharose CL-6B (AN-Sepharose) for use as a new ligand of mixed-mode chromatography. Adsorption equilibria of immunoglobulin G (IgG) and bovine serum albumin (BSA) to AN-Sepharose were studied at extensive pH values (4.0-8.8) and salt
Dong-Sheng Chai et al.
Journal of separation science, 37(23), 3461-3472 (2014-09-19)
Efficient loading of immunoglobulin G in mixed-mode chromatography is often a serious bottleneck in the chromatographic purification of immunoglobulin G. In this work, a mixed-mode ligand, 4-(1H-imidazol-1-yl) aniline, was coupled to Sepharose Fast Flow to fabricate AN SepFF adsorbents with
Lin-Ling Yu et al.
Journal of separation science, 34(21), 2950-2959 (2011-09-29)
In the current research, a series of dextran-grafted adsorbents were prepared using sulfopropyl and 4-(1H-imidazol-1-yl) aniline as chromatographic ligands for ion-exchange (IEC) and mixed-mode chromatography (MMC) to respectively investigate the influence of dextran layer on adsorption of γ-globulin. Experimental evidences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service