Skip to Content
Merck
All Photos(1)

Key Documents

440981

Sigma-Aldrich

(R)-(−)-3-Chloro-1,2-propanediol

97%

Synonym(s):

(R)-α-Glycerol chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
110.54
Beilstein:
2202928
MDL number:
UNSPSC Code:
12352000

vapor pressure

0.04 mmHg ( 25 °C)

Assay

97%

optical activity

[α]20/D −0.9°, neat

optical purity

ee: 97% (GLC)

refractive index

n20/D 1.48 (lit.)

bp

213 °C (lit.)

density

1.321 g/mL at 20 °C (lit.)

SMILES string

OC[C@@H](O)CCl

InChI

1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2/t3-/m0/s1

InChI key

SSZWWUDQMAHNAQ-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of (S)-(-)-pindolol and chuangxinmycin.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ishibashi, H. et al.
Chemical & Pharmaceutical Bulletin, 42, 271-271 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service