Skip to Content
Merck
All Photos(1)

Key Documents

440868

Sigma-Aldrich

(R)-(+)-3-Chlorostyrene oxide

98%

Synonym(s):

(R)-(+)-(3-Chlorophenyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO
CAS Number:
Molecular Weight:
154.59
MDL number:
UNSPSC Code:
12352000

Assay

98%

optical activity

[α]20/D +21°, neat

refractive index

n20/D 1.551 (lit.)

bp

67-68 °C/1 mmHg (lit.)

density

1.214 g/mL at 25 °C (lit.)

SMILES string

Clc1cccc(c1)[C@@H]2CO2

InChI

1S/C8H7ClO/c9-7-3-1-2-6(4-7)8-5-10-8/h1-4,8H,5H2/t8-/m0/s1

InChI key

YVMKRPGFBQGEBF-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Key chiral synthon for the preparation of the potent antidiabetic and antiobesity agent CL-316,243, a benzodioxole-containing phenethanolamine. Ring opening of this chiral epoxide with amines produces amino alcohols which have been used as β-adrenergic agonists.
Reactant involved in:
  • Diastereoselective Wadsworth-Emmons cyclopropanation for synthesis of quaternaty cyclopropyl esters
  • Synthesis of phenylethanolamine derivatives as β3-adrenergic receptor agonists
  • Stereoselective ring-opening for synthesis of β-aryloxy-substituted alcohols and amines
  • Synthesis of Biphenyl benzoic acid derivatives as β3-adrenergic receptor agonists
  • Synthesis of (S)-2-, 3-, and 4-chlorostyrene oxides with epoxide hydrolase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disodium (R,R)-5-[2-[[2-(3-chlorophenyl)-2-hydroxyethyl]-amino] propyl]-1,3-benzodioxole-2,2-dicarboxylate (CL 316,243). A potent beta-adrenergic agonist virtually specific for beta 3 receptors. A promising antidiabetic and antiobesity agent.
J D Bloom et al.
Journal of medicinal chemistry, 35(16), 3081-3084 (1992-08-07)
D. Badone et al.
Bioorganic & Medicinal Chemistry Letters, 16, 1921-1921 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service