Skip to Content
Merck
All Photos(2)

Key Documents

440825

Sigma-Aldrich

Tris[3-(trimethoxysilyl)propyl] isocyanurate

technical grade

Synonym(s):

1,3,5-N-Tris[3-(trimethoxysilyl)propyl] isocyanurate, 1,3,5-Tris(3-trimethoxysilylpropyl)-1,3,5-triazinane-2,4,6-trione, Tris(3-trimethoxysilylpropyl)cyclotriisocyanurate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$167.00

$167.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$167.00

About This Item

Empirical Formula (Hill Notation):
C21H45N3O12Si3
CAS Number:
Molecular Weight:
615.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$167.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

technical grade

form

liquid

refractive index

n20/D 1.461 (lit.)

viscosity

350 cSt(lit.)

bp

250 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCN1C(=O)N(CCC[Si](OC)(OC)OC)C(=O)N(CCC[Si](OC)(OC)OC)C1=O)(OC)OC

InChI

1S/C21H45N3O12Si3/c1-28-37(29-2,30-3)16-10-13-22-19(25)23(14-11-17-38(31-4,32-5)33-6)21(27)24(20(22)26)15-12-18-39(34-7,35-8)36-9/h10-18H2,1-9H3

InChI key

QWOVEJBDMKHZQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris[3-(trimethoxysilyl)propyl] isocyanurate (TTPI) can be used in the preparation of:
  • Pd nanoparticle and single-walled carbon nanotube hybrid materials.[1]
  • Periodic mesoporous organosilicas exhibiting absorption properties for heavy metal ions like mercury.[2]
  • Mesoporous organosilica−alumina composites with an affinity for CO2 gas at elevated temperatures.[3]
  • Isocyanurate-containing ordered mesoporous organosilicas using block copolymer.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.3 °F - closed cup

Flash Point(C)

93.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous isocyanurate-containing organosilica-alumina composites and their thermal treatment in nitrogen for carbon dioxide sorption at elevated temperatures
Gunathilake C, et al.
Journal of Material Chemistry A, 1(28), 8244-8252 (2013)
Synthesis and characterization of novel hybrids of tris [3-(trimethoxysilyl) propyl] isocyanurate (TTPI) capped palladium nanoparticles and single-walled carbon nanotubes
Hemraj-Benny T, et al.
Silicon, 3(2), 97-101 (2011)
Cage-like mesoporous organosilicas with isocyanurate bridging groups synthesized by soft templating with poly (ethylene oxide)-poly (butylene oxide)-poly (ethylene oxide) block copolymer
Grudzien RM, et al.
Journal of Colloid and Interface Science, 333(1), 354-362 (2009)
Bifunctional periodic mesoporous organosilica with large heterocyclic bridging groups and mercaptopropyl ligands
Olkhovyk O, et al.
Journal of Materials Chemistry, 15(15), 1517-1519 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service