Skip to Content
Merck
All Photos(2)

Key Documents

439983

Sigma-Aldrich

Methyl 2-isocyanatobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNC6H4CO2CH3
CAS Number:
Molecular Weight:
177.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

101 °C/2 mmHg (lit.)

mp

45-49 °C (lit.)

functional group

ester
isocyanate

SMILES string

COC(=O)c1ccccc1N=C=O

InChI

1S/C9H7NO3/c1-13-9(12)7-4-2-3-5-8(7)10-6-11/h2-5H,1H3

InChI key

ATFKBWNEVZUSKC-UHFFFAOYSA-N

Related Categories

Application

Methyl 2-isocyanatobenzoate may be used for the following syntheses:
  • 3H-quinazolin-4-ones
  • A1120, a high affinity (Ki = 8.3nM) non-retinoid ligand for the Retinol-binding protein
  • trans- and cis-N-phenyl-N′-(4-phenoxy)cyclohexylureas

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander V Lygin et al.
Organic letters, 11(2), 389-392 (2008-12-23)
ortho-Lithiophenyl isocyanide has been generated from ortho-bromophenyl isocyanide and successfully employed toward the synthesis of 2-substituted phenyl isocyanides as well as 2,3-disubstituted 3H-quinazoline-4-ones and 3H-quinazolin-4-thiones.
Chemistry of 4H-3, 1-Benzoxazin-4-ones.
El-Mekabaty A.
International Journal of Modern Organic Chemistry, 2(2), 81-121 (2013)
Alykhan Motani et al.
The Journal of biological chemistry, 284(12), 7673-7680 (2009-01-17)
Retinol-binding protein 4 (RBP4) transports retinol from the liver to extrahepatic tissues, and RBP4 lowering is reported to improve insulin sensitivity in mice. We have identified A1120, a high affinity (K(i) = 8.3 nm) non-retinoid ligand for RBP4, which disrupts
Ting Chen et al.
Journal of medicinal chemistry, 56(23), 9457-9470 (2013-11-23)
Heme-regulated inhibitor kinase (HRI), a eukaryotic translation initiation factor 2 alpha (eIF2α) kinase, plays critical roles in cell proliferation, differentiation, and adaptation to cytoplasmic stress. HRI is also a critical modifier of hemoglobin disorders such as β-thalassemia. We previously identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service