Skip to Content
Merck
All Photos(1)

Key Documents

439509

Sigma-Aldrich

3-Acetyl-2,5-dichlorothiophene

98%

Synonym(s):

2,5-Dichloro-3-thienyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2OS
CAS Number:
Molecular Weight:
195.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

37-40 °C (lit.)

SMILES string

CC(=O)c1cc(Cl)sc1Cl

InChI

1S/C6H4Cl2OS/c1-3(9)4-2-5(7)10-6(4)8/h2H,1H3

InChI key

GYFDNIRENHZKGR-UHFFFAOYSA-N

Application

3-Acetyl-2,5-dichlorothiophene may be used in the following syntheses:
  • substituted chalcones[1]
  • AL-4623A and AL-4862 (brinzolamide), inhibitors of topical carbonic anhydrase[2]
  • 2,5-dichlorothiophene-3-carboxylic acid, which is the starting reagent required for the synthesis of 2,5-dichloro-N-(substituted amino-carbonothioyl)thiophene-3-carboxamides and 2-(substituted amino)-4H-thieno[3,2-e]-1,3-thiazin-4-ones[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thienopyridone Antibacterials. Part IV [1]. Synthesis of Some N (7)-Heteroaryl-4-oxothieno [2, 3-b] pyridine-5-carboxylic Acids and Esters.
Sweidan NI, et al.
Letters in Organic Chemistry, 7(1), 79-84 (2010)
Enantioselective Synthesis of Brinzolamide (AL-4862), a New Topical Carbonic Anhydrase Inhibitor. The ?DCAT Route? to Thiophenesulfonamides.
Conrow RE, et al.
Organic Process Research & Development, 3(2), 114-120 (1999)
Thienopyridone Antibacterials V. Synthesis of Some N (7)-Azacyclohexyl-4-oxothieno [2, 3-b] pyridine-5-carboxylic Acids and Related Congeners.
Al-Jaber HI, et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 19(11), 1625-1625 (2009)
Facile synthesis of 2-(substituted amino)-4H-thieno [3, 2-e]-1, 3-thiazin-4-ones.
Abu-El-Halawa R, et al.
Monatshefte fur Chemie / Chemical Monthly, 139(10), 1251-1255 (2008)
V Tomar et al.
Bioorganic & medicinal chemistry letters, 17(19), 5321-5324 (2007-08-28)
Two new series of chalcones have been synthesized by reacting 1-(4-piperazin-1-yl-phenyl)ethanone and 1-(2,5-dichloro-3-thienyl)-1-ethanone with different substituted benzaldehydes in turn by Claisen-Schmidt condensation. The compounds have been characterized by IR, (1)H NMR spectral and microanalysis data. All the synthesized compounds have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service