Skip to Content
Merck
All Photos(2)

Key Documents

438804

Sigma-Aldrich

2-Nitrotoluene

≥99%

Synonym(s):

1-Methyl-2-nitrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$56.20

$56.20


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$56.20

About This Item

Linear Formula:
CH3C6H4NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
1907580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥99%

$56.20


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.546 (lit.)

bp

225 °C (lit.)

mp

−4-−3 °C (lit.)

density

1.163 g/mL at 25 °C (lit.)

functional group

nitro

SMILES string

Cc1ccccc1[N+]([O-])=O

InChI

1S/C7H7NO2/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3

InChI key

PLAZTCDQAHEYBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Nitrotoluene is a nitroaromatic compound. Multi-functional magnetic resins having high surface area and ion exchange efficiency (GMA30-1 and GMA30-2) were developed and were tested for the desorption of 2-nitrotolene.[1] Its dioxygenation by nitrobenzene dioxygenase has been studied to evaluate the isotope fractionation.[2] It has been reported as nitroaromatic explosive. Terbium-based magnetic metal-organic framework (MOF) nanospheres have been fabricated, which can detect the trace quantities of 2-nitrotoluene.[3]

Application

2-Nitrotoluene may be employed as nitrogen supplement in the culture medium of Pseudomonas sp. strain ClS1.[4] It may be employed as carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.[5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 1B - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jing-Jing Qian et al.
Dalton transactions (Cambridge, England : 2003), 43(10), 3978-3983 (2014-01-24)
In this work, we present novel kinds of Fe3O4@Tb-BTC magnetic metal-organic framework (MOF) nanospheres which possess both magnetic characteristics and fluorescent properties using a layer by layer assembly technique. The structure and morphology of the as-prepared Fe3O4@Tb-BTC were systematically characterized
Sarah G Pati et al.
Environmental science & technology, 48(18), 10750-10759 (2014-08-08)
Oxygenation of aromatic rings is a frequent initial step in the biodegradation of persistent contaminants, and the accompanying isotope fractionation is increasingly used to assess the extent of transformation in the environment. Here, we systematically investigated the dioxygenation of two
Wei Wang et al.
Journal of hazardous materials, 294, 158-167 (2015-04-14)
Two novel multi-functional magnetic resins named GMA30-1 and GMA30-2 were fabricated and investigated for the removal of 4-nitrotoluene-2-sulfonic acid (NTS) and 2-nitrotoluene (o-MNT). Strong base resin (GMA30-1) and weak base resin (GMA30-2) possess large surface area of 718m(2)/g and 559m(2)/g
E Duque et al.
Journal of bacteriology, 175(8), 2278-2283 (1993-04-01)
A bacterium, Pseudomonas sp. strain C1S1, able to grow on 2,4,6-trinitrotoluene (TNT), 2,4- and 2,6-dinitrotoluene, and 2-nitrotoluene as N sources, was isolated. The bacterium grew at 30 degrees C with fructose as a C source and accumulated nitrite. Through batch
D An et al.
Journal of bacteriology, 176(24), 7462-7467 (1994-12-01)
Pseudomonas sp. strain JS42 utilizes 2-nitrotoluene (2NT) as the sole source of carbon and energy for growth. Intact cells catalyze the oxidation of 2NT to 3-methylcatechol and nitrite in a reaction that requires molecular oxygen. Cell extracts oxidized 2NT to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service