Skip to Content
Merck
All Photos(2)

Key Documents

437980

Sigma-Aldrich

Zinc methacrylate

Synonym(s):

Methacrylic acid zinc salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$69.40

$69.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$69.40

About This Item

Linear Formula:
[H2C=C(CH3)CO2]2Zn
CAS Number:
Molecular Weight:
235.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

$69.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

form

powder

mp

229-232 °C (lit.)

SMILES string

CC(=C)C(=O)O[Zn]OC(=O)C(C)=C

InChI

1S/2C4H6O2.Zn/c2*1-3(2)4(5)6;/h2*1H2,2H3,(H,5,6);/q;;+2/p-2

InChI key

PIMBTRGLTHJJRV-UHFFFAOYSA-L

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service