Skip to Content
Merck
All Photos(2)

Key Documents

437220

Sigma-Aldrich

Poly(ethylene-co-vinyl acetate)

vinyl acetate 25 wt. %, melt index 19 g/10 min (190°C/2.16 kg), contains 200-900 ppm BHT as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)m[CH2CH(OCOCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

autoignition temp.

500 °F

melt index

19 g/10 min (190°C/2.16 kg)

contains

200-900 ppm BHT as inhibitor

composition

vinyl acetate, 25 wt. %

hardness

28 (Shore D, ASTM D 2240)

mp

75 °C

transition temp

softening point 46 °C (Vicat, ASTM D 1525)

density

0.948 g/mL at 25 °C

SMILES string

C=C.CC(=O)OC=C

InChI

1S/C4H6O2.C2H4/c1-2-3-4(5)6;1-2/h2H,1,3H2,(H,5,6);1-2H2

InChI key

DQXBYHZEEUGOBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Flexible tubing, color concentrates, gaskets and molded parts for autos, plastic lenses and pumps.

Features and Benefits

Thermoplastic, resilient, tough and resistant to ozone and environmental stress cracking. Inherently flexible, requiring no plasticizers.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eva C González Díaz et al.
Tissue engineering. Part A, 24(13-14), 1148-1156 (2018-01-26)
Synthetic biomaterials that create a dynamic calcium (Ca2+)-, phosphate (PO43-) ion-, and calcium phosphate (CaP)-rich microenvironment, similar to that found in native bone tissue, have been shown to promote osteogenic commitment of stem cells in vitro and in vivo. The
Ethylene vinyl acetate (foam): an inexpensive and useful tool for teaching suture techniques in dermatologic surgery.
Daniela Gutiérrez-Mendoza et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 37(9), 1353-1357 (2011-05-28)
Martin G Jorgensen et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 68(7), 845-852 (2012-11-02)
Older adults show increased risk of falling and major risk factors include impaired lower extremity muscle strength and postural balance. However, the potential positive effect of biofeedback-based Nintendo Wii training on muscle strength and postural balance in older adults is
Rafael Denadai et al.
Acta cirurgica brasileira, 27(3), 271-278 (2012-03-31)
To assess the acquisition of suture skills by training on ethylene-vinyl acetate bench model in novice medical students. Sixteen medical students without previous surgery experience (novices) were randomly divided into two groups. During one hour group A trained sutures on
Neide Pena Coto et al.
Dental traumatology : official publication of International Association for Dental Traumatology, 28(2), 108-113 (2011-07-28)
There has been a significant increase in the number of facial fractures stemming from sport activities in recent years, with the nasal bone one of the most affected structures. Researchers recommend the use of a nose protector, but there is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service