Skip to Content
Merck
All Photos(1)

Key Documents

436763

Sigma-Aldrich

4-Chloro-2-fluorobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CO2H
CAS Number:
Molecular Weight:
174.56
Beilstein:
973358
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

204-208 °C (lit.)

SMILES string

OC(=O)c1ccc(Cl)cc1F

InChI

1S/C7H4ClFO2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)

InChI key

ZLPXBWMVZANJJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-2-fluorobenzoic acid is a halogen benzoic acid. It is a non-liquid crystalline carboxylic acid derivative. It has been reported to form hydrogen bonded complexes with benzoylhydrazine-based azobenzene compound.[1] Octanol/water partition coefficient Kow of 4-chloro-2-fluorobenzoic acid has been determined by shake-flask method.[2]

Application

4-Chloro-2-fluorobenzoic acid may be used in the following studies:
  • As starting reagent for the synthesis of furosemide.[3]
  • Synthesis of 4′-chloro-2′-fluoroacetophenone.[4]
  • Synthesis of novel herbicidal isoxazolecarboxamides.[5]
  • Preparation of potential liquid crystals.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Octanol/Water partition coefficient of substituted benzene derivatives containing halogens and carboxyls: Determination using the shake-flask method and estimation using the fragment method.
Qiao Y, et al.
Journal of Chemical and Engineering Data, 53(1), 280-282 (2007)
Helvetica Chimica Acta, 68, 1444-1444 (1985)
Selective lithiation/carbonation of polyhalobenzenes: an improved synthesis of furosemide-7-14C.
Perry CW, et al.
The Journal of Organic Chemistry, 43(22), 4391-4391 (1978)
A New and Versatile Synthesis of Thioflavanones from 2-Fluorobenzoic Acids.
Lee JI.
J. Korean Chem. Soc., 58(4) (2014)
Self-assembled 1, 2-bis [4-(4-(10-decyloxy) phenylazo)] benzoylhydrazine dimer and its hydrogen-bonded complexes.
Yeap G-Y, et al.
Supramolecular Chemistry, 25(7), 424-431 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service