Skip to Content
Merck
All Photos(2)

Key Documents

434566

Sigma-Aldrich

Poly(4-styrenesulfonic acid-co-maleic acid) sodium salt

average Mw ~20,000, powder

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$69.80

$69.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$69.80

About This Item

Linear Formula:
[CH2CH(C6H4SO3R)]x[CH(CO2R)CH(CO2R)]y, R=H or Na
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$69.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average Mw ~20,000

viscosity

30 cP, 25 wt. % in H2O(lit.)

solubility

H2O: soluble

density

0.801 g/mL at 25 °C

SMILES string

[Na+].O2C(=O)C=CC2=O.c1(ccccc1)C=C

InChI

1S/C8H8.C4H2O3.Na/c1-2-8-6-4-3-5-7-8;5-3-1-2-4(6)7-3;/h2-7H,1H2;1-2H;/q;;+1

InChI key

AGYUYWQDVRLUBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3:1 4-styrenesulfonic acid:maleic acid mole ratio

Application

Boiler additive to prevent and remove sludge. Crystal modifier for calcium salts and excellent dispersant for iron oxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik C Gunther et al.
Cell reports, 26(1), 145-158 (2019-01-04)
Cellular prion protein (PrPC) binds the scrapie conformation of PrP (PrPSc) and oligomeric β-amyloid peptide (Aβo) to mediate transmissible spongiform encephalopathy (TSE) and Alzheimer's disease (AD), respectively. We conducted cellular and biochemical screens for compounds blocking PrPC interaction with Aβo.

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service