Skip to Content
Merck
All Photos(1)

Key Documents

434507

Sigma-Aldrich

Poly(methacrylic acid, sodium salt) solution

average Mn ~5,400, average Mw ~9,500 by GPC, 30 wt. % in H2O

Synonym(s):

Poly(methacrylic acid) sodium salt, Sodium polymethacrylate, Sodium polymethacrylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$154.00

About This Item

Linear Formula:
[CH2C(CH3)(CO2Na)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

mol wt

average Mn ~5,400
average Mw ~9,500 by GPC

concentration

30 wt. % in H2O

pH

8-9

viscosity

110-375 cP(25 °C, Brookfield)(lit.)

density

1.18 g/mL at 25 °C

SMILES string

CC(=C)C(=O)O[Na]

InChI

1S/C4H6O2.Na/c1-3(2)4(5)6;/h1H2,2H3,(H,5,6);/q;+1/p-1

InChI key

SONHXMAHPHADTF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Water treatment agent for cooling towers, boilers and heat exchangers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Stefaan J Soenen et al.
ACS nano, 6(7), 5767-5783 (2012-06-05)
The in vitro labeling of therapeutic cells with nanoparticles (NPs) is becoming more and more common, but concerns about the possible effects of the NPs on the cultured cells are also increasing. In the present work, we evaluate the effects
Wataru Takada et al.
Analytical biochemistry, 435(2), 123-130 (2013-01-22)
Glycosaminoglycans (GAGs) take part in numerous biological processes by binding to protein molecules and functionally regulating protein-ligand interactions; therefore, molecular interactions of GAGs have been studied by several methods, including surface plasmon resonance, enzyme-linked immunosorbent assays (ELISAs), and GAG microarrays.
Pavel Jandera et al.
Journal of chromatography. A, 1274, 97-106 (2013-01-01)
We synthesized 8 polymethacrylate monolithic capillary columns using laurylmethacrylate functional monomer and various cross-linking monomers differing in the polarity and size. The efficiency of monolithic columns for low-molecular compounds significantly improved with increasing number of repeat non-polar methylene groups in
Ania Servant et al.
Advanced healthcare materials, 2(6), 806-811 (2012-11-28)
Drug release triggered by an external non-invasive stimulus is of great interest for the development of new drug delivery systems. The preparation of an electroresponsive multiwalled carbon nanotube/poly(methylacrylic acid) (MWNT/PMAA)-based hybrid material is reported. The hydrogel hybrids achieve a controlled
Alireza Shalviri et al.
Colloids and surfaces. B, Biointerfaces, 101, 405-413 (2012-09-27)
This work focused on the design of new pH-responsive nanoparticles for controlled delivery of anticancer drug doxorubicin (Dox). Nanoparticles of poly(methacrylic acid)-polysorbate 80-grafted starch (PMAA-PS 80-g-St) were synthesized by using a one-pot method that enabled simultaneous grafting of PMAA and

Questions

  1. Is the polymer atactic?

    1 answer
    1. Tacticity is not specified for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service