Skip to Content
Merck
All Photos(1)

Key Documents

433853

Sigma-Aldrich

5-Chloro-3-methyl-1-phenyl-4-pyrazolecarboxaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9ClN2O
CAS Number:
Molecular Weight:
220.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

145-148 °C (lit.)

functional group

aldehyde
chloro

SMILES string

Cc1nn(c(Cl)c1C=O)-c2ccccc2

InChI

1S/C11H9ClN2O/c1-8-10(7-15)11(12)14(13-8)9-5-3-2-4-6-9/h2-7H,1H3

InChI key

DKZPJLZXLKAMDO-UHFFFAOYSA-N

General description

5-Chloro-3-methyl-1-phenyl-4-pyrazolecarboxaldehyde (5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde) can be synthesized from 3-methyl-1-phenyl-1Hpyrazol-5(4H)-one under Vilsmeier-Haack reaction conditions. Knoevenagel condensation of 5-chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde with ethylcyanoacetate at 0°C has been reported to afford ethyl-2-cyano-3-(5-chloro-3-methyl-1-phenyl-1H-pyrazole-4-yl)propionate. It participates in the synthesis of 2-((5-phenoxy-3-methyl-1-phenyl-1H-pyrazol- 4-yl)methylene)hydrazinecarbothiomide derivatives with a potential anticonvulsant property.

Application

5-Chloro-3-methyl-1-phenyl-4-pyrazolecarboxaldehyde (5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde) may be employed as starting material for the following syntheses:
  • 5-chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde hydrazone
  • N1-((5-chloro-3-methyl-1-phenyl-1H-pyrazol-4-yl)methylene)thiosemicarbazone
  • pyrazole derivatives, which were pharmacologically evaluated for analgesic (tail flick) and anti-inflammatory (based on carrageenan-induced paw edema) activities
  • N′-[(5-chloro-3-methyl-1-phenyl-1H-pyrazol-4- yl)methylene] 2/4-substituted hydrazides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New pyrazole derivatives of potential biological activity.
Farghaly A-R, et al.
ARKIVOC (Gainesville, FL, United States), 7, 228-241 (2012)
Convenient Synthesis and Characterization of Pyrazolones and Schiff Bases.
Girisha KS and Kalluraya B.
Synthetic Communications, 42(20), 3097-3102 (2012)
Design and Synthesis of Pyrazolyl Thiosemicarbazones as New Anticonvulsants.
Deng XQ and Song MX.
Bull. Korean Chem. Soc., 35(9), 2733-2733 (2014)
Pharmacological screening for anti-inflammatory, analgesic activity of pyrazolyl derivatives along with molecular docking studies.
Kaushik D, et al.
Medicinal Chemistry Research, 21(11), 3646-3655 (2012)
Darpan Kaushik et al.
European journal of medicinal chemistry, 45(9), 3943-3949 (2010-06-25)
A series of N'-[(5-chloro-3-methyl-1-phenyl-1H-pyrazol-4-yl)methylene] 2/4-substituted hydrazides were synthesized using appropriate synthetic route and characterized by elemental analysis and spectral data. The anticonvulsant activity of some of the synthesized compounds were evaluated against maximal electroshock induced seizure (MES) and subcutaneous pentylenetetrazol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service