All Photos(1)
About This Item
Linear Formula:
LiOCl
CAS Number:
Molecular Weight:
58.39
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
composition
available chlorine, >70%
SMILES string
[Li+].[O-]Cl
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Ox. Sol. 2 - Skin Corr. 1B
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N L Davies
Journal of the South African Veterinary Association, 62(3), 140-142 (1991-09-01)
Two cases of lithium toxicity are reported on in dogs having had lithium hypochlorite chlorinated water as their sole source of drinking water. Clinical signs in one dog included polyuria, polydipsia, loss of body mass; dehydration, diarrhoea and general weakness
M L Weiner et al.
Toxicology, 65(1-2), 1-22 (1990-12-17)
Lithium hypochlorite (LiOCl), the pool and spa sanitizer/algicide, was evaluated for genotoxicity in a battery of studies designed to evaluate potential mutagenicity, DNA damage and chromosome aberrations. LiOCl was not mutagenic in the Ames test when tested in Salmonella typhimurium
Tzu-Chao Lin et al.
Epilepsy & behavior : E&B, 14(2), 316-323 (2009-01-08)
Neonatal seizures may alter the developing neurocircuitry and cause behavioral abnormalities in adulthood. We found that rats previously subjected to lithium-pilocarpine (LiPC)-induced neonatal status epilepticus (NeoSE) exhibited enhanced behavioral sensitization to methamphetamine (MA) in adolescence. Neurochemically, dopamine (DA) and metabolites
Keith C Donavan et al.
Langmuir : the ACS journal of surfaces and colloids, 28(34), 12581-12587 (2012-08-04)
Virus-poly(3,4-ethylenedioxythiophene) (virus-PEDOT) biocomposite films are prepared by electropolymerizing 3,4-ethylenedioxythiophene (EDOT) in aqueous electrolytes containing 12 mM LiClO(4) and the bacteriophage M13. The concentration of virus in these solutions, [virus](soln), is varied from 3 to 15 nM. A quartz crystal microbalance
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service