Skip to Content
Merck
All Photos(3)

Key Documents

432490

Sigma-Aldrich

Polystyrene-block-polybutadiene

30 wt.% styrene

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$81.70
1 KG
$138.00

$81.70

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$81.70
1 KG
$138.00

About This Item

Linear Formula:
[CH2CH(C6H5)]x (CH2CH=CHCH2)y [CH2CH(C6H5)]x
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$81.70

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

form

beads

melt index

10 g/10 min (200°C/5.0kg)

contains

phenolic as antioxidant

hardness

61 (Shore A)

viscosity

8.5 poise, 25 wt. % in toluene(25 °C, Brookfield)(lit.)

solubility

solvents with solubility parameters between 7.7 and 9.4: soluble

elongation

ASTM D 412 - 600% (ultimate)

Application

Component of hot melt adhesives, sealants, asphalt and oil gels. Modifier for polymers, thermosets and general rubber compounding. Direct molded or extruded auto parts, sporting goods, footware and film.

Features and Benefits

High strength and low viscosity for easy melt and solution processing. High temperature degradation results in cross-linking. Flexible to -65°C. Excellent electrical insulator. Resists acids and alkali.

Physical form

Thermoplastic elastomer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sneha B Chakrapani et al.
The Analyst, 144(5), 1679-1686 (2019-01-19)
The accurate characterization of relative composition in multicomponent polymer systems such as statistical copolymers, block copolymers, and polymer blends is critical to understanding and predicting their behavior. Typically, polymer compositional analysis is performed using 1H Nuclear Magnetic Resonance (NMR) Spectroscopy

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Can I know the average molecular weight information of the polymer?

    1 answer
    1. The molecular weight is approximately 153-185 kda.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service