Skip to Content
Merck
All Photos(1)

Key Documents

431893

Sigma-Aldrich

(S)-(+)-1,1′-Bi-2-naphthol bis(trifluoromethanesulfonate)

97%

Synonym(s):

(S)-1,1′-Bi(2-naphthol) bis(trifluoromethanesulfonate), (S)-1,1′-Binaphthalene-2,2′-diyl bis(trifluoromethanesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3C10H6-)2
CAS Number:
Molecular Weight:
550.45
Beilstein:
3579131
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]21/D +148°, c = 1 in chloroform

mp

83-85 °C (lit.)

InChI

1S/C22H12F6O6S2/c23-21(24,25)35(29,30)33-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)34-36(31,32)22(26,27)28/h1-12H

InChI key

OYJLCOSEYYZULE-UHFFFAOYSA-N

Application

Used as an intermediate in the preparation of chiral phosphine ligands for asymmetric synthesis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 1945-1945 (1993)
Acta Chemica Scandinavica, 50, 259-259 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service