Skip to Content
Merck
All Photos(1)

Key Documents

431850

Sigma-Aldrich

Europium(II) chloride

99.99% trace metals basis

Synonym(s):

Dichloroeuropium, Europium chloride, Europous chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$275.00
5 G
$950.00

$275.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$275.00
5 G
$950.00

About This Item

Linear Formula:
EuCl2
CAS Number:
Molecular Weight:
222.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

$275.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Assay

99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: europium

impurities

<0.1% Zn

SMILES string

Cl[Eu]Cl

InChI

1S/2ClH.Eu/h2*1H;/q;;+2/p-2

InChI key

BMANZYKPCBPZFG-UHFFFAOYSA-L

General description

Europium (II) chloride is awhite to pale pink solid with orthorhombic crystal structure. It is a stablematerial that is hygroscopic in nature. It can be prepared simply by reducingEuropium (III) chloride with hydrogen gas at high temperatures. It has atunable luminescence.

Application

Europium(II) chloride can be used as:
  • A dopant to prepare phosphor materials for energy-efficient solid-state lighting.[1]
  • A pressure-sensor material.[2]
  • A suitable precursor for the synthesis of Europium(ii) compounds coordination polymers with 2,2′-bipyrimidine-mediated ferromagnetic interactions.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Europium (II) compounds: simple synthesis of a molecular complex in water and coordination polymers with 2, 2?-bipyrimidine-mediated ferromagnetic interactions
Gael Zucchi, et al
Chemical Communications (Cambridge, England), 46, 9143-9145 (2010)
EuCl2 Powder as a Pressure-Memory Material
Takamichi Kobayashi
Japanese Journal of Applied Physics, 49, 088001-088001 (2010)
Europium-activated orthophosphate phosphors for energy-efficient solid-state lighting: a review
KN Shinde and SJ Dhoble
Critical Reviews in Solid State and Materials Sciences, 39, 459-479 (2014)

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service