Skip to Content
Merck
All Photos(1)

Documents

431249

Sigma-Aldrich

Trimethyl phosphite

≥99.999% trace metals basis

Synonym(s):

Methyl phosphite, P(OMe)3, Trimethoxyphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3P
CAS Number:
Molecular Weight:
124.08
Beilstein:
956570
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.3 (vs air)

vapor pressure

17 mmHg ( 20 °C)

Assay

≥99.999% trace metals basis

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.408 (lit.)

bp

111-112 °C (lit.)

mp

−78 °C (lit.)

density

1.052 g/mL at 25 °C (lit.)

SMILES string

COP(OC)OC

InChI

1S/C3H9O3P/c1-4-7(5-2)6-3/h1-3H3

InChI key

CYTQBVOFDCPGCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 15 Phosphorus

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian-lin Zhang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(11), 1334-1337 (2005-03-15)
The reactions of unsaturated chloride polyether polyol with trimethyl phosphite have been studied by FTIR, which was prepared by copolymerization of ethanediol, allyl glycidyl ether and epoxychloropropane. The experimental results showed that the reaction included ester-exchange reaction, ester-exchange polymerization and
N Ramanathan et al.
The journal of physical chemistry. A, 116(49), 12014-12023 (2012-11-21)
Trimethyl phosphite (TMPhite) and hydrogen chloride (HCl), when separately codeposited in a N(2) matrix, yielded a hydrogen bonded adduct, which was evidenced by shifts in the vibrational frequencies of the TMPhite and HCl submolecules. The structure and energy of the
D Vaughan Griffiths et al.
Organic & biomolecular chemistry, 6(3), 577-585 (2008-01-26)
Dialkyl heteroaroylphosphonates based on thiophene, pyrrole or furan have been prepared and their reactions with trimethyl phosphite investigated. Deoxygenation of the carbonyl groups in these heteroaroylphosphonates occurs to give carbene intermediates, which then undergo further reaction. In the case of
M A Mehlman et al.
Toxicology and applied pharmacology, 72(1), 119-123 (1984-01-01)
Trimethyl phosphite (TMP) is an organophosphorus alkylating agent used primarily in the synthesis of organophosphate compounds. To evaluate teratogenic potential, TMP was administered by gavage to pregnant rats at rates of 16, 49, or 164 mg/kg/day, on gestation Days 6
Bangwei Ding et al.
Journal of the American Chemical Society, 125(11), 3248-3259 (2003-03-13)
Trimethyl phosphite, (MeO)(3)P, is introduced as an efficient and selective trap in oxiranylcarbinyl radical (2) systems, formed from haloepoxides 8-13 under thermal AIBN/n-Bu(3)SnH conditions at about 80 degrees C. Initially, the transformations of 8-13, in the absence of phosphite, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service