Skip to Content
Merck
All Photos(1)

Key Documents

429910

Sigma-Aldrich

2-Fluoro-6-iodobenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$261.00

$261.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$261.00

About This Item

Linear Formula:
FC6H3(I)CO2H
CAS Number:
Molecular Weight:
266.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$261.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

123-126 °C (lit.)

functional group

carboxylic acid
fluoro
iodo

SMILES string

OC(=O)c1c(F)cccc1I

InChI

1S/C7H4FIO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H,10,11)

InChI key

CYCXAPWOBWWNRK-UHFFFAOYSA-N

General description

2-Fluoro-6-iodobenzoic acid is loose white crystal. It can be synthesized by using 2-amino-6-fluorobenzoic acid as the starting material.[1]

Application

2-Fluoro-6-iodobenzoic acid may be used for the synthesis of fluoro-substituted benzoyl chlorides.[2] It may be used for the one-pot regioselective synthesis of isocoumarins.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-fluoro-6-iodobenzoic acid.
Zhao H-Y, et al.
Chemical Reagents, 11, 028-028 (2010)
Peri fluoro steric effects: syntheses and comparative acid-catalyzed isomerization of the 8-, 9-, and 11-fluoro-1, 2, 3, 4-tetrahydro-7, 12-dimethylbenz [a] anthracenes to exo methylene tautomers.
Witiak DT, et al.
The Journal of Organic Chemistry, 53(2), 345-352 (1988)
Regioselective One-Pot Synthesis of Isocoumarins and Phthalides from 2-Iodobenzoic Acids and Alkynes by Temperature Control.
Kumar MR, et al.
Advanced Synthesis & Catalysis, 355(16), 3221-3230 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service