Skip to Content
Merck
All Photos(1)

Key Documents

428558

Sigma-Aldrich

5-(4-Nitrophenyl)-2-furoic acid

96%

Synonym(s):

5-(4-Nitrophenyl)-2-furancarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$74.80

$74.80


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$74.80

About This Item

Empirical Formula (Hill Notation):
C11H7NO5
CAS Number:
Molecular Weight:
233.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.80


Estimated to ship onMay 30, 2025


Request a Bulk Order

Assay

96%

mp

255-257 °C (dec.) (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1ccc(o1)-c2ccc(cc2)[N+]([O-])=O

InChI

1S/C11H7NO5/c13-11(14)10-6-5-9(17-10)7-1-3-8(4-2-7)12(15)16/h1-6H,(H,13,14)

InChI key

FXZTYDPWMQBTDH-UHFFFAOYSA-N

General description

5-(4-Nitrophenyl)-2-furoic acid (5-(p-nitrophenyl)-2-furoic acid) is a 5-aryl-2-furoic acid. It is a dantrolene metabolite and its presence in plasma and urine has been determined by HPLC.[1] It was reported to be the only product formed during the reduction of 5-(4-nitrophenyl)-2-furaldehyde by modified Cannizzaro reaction.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of a dantrolene metabolite, 5-(p-nitrophenyl)-2-furoic acid, in plasma and urine by high-performance liquid chromatography.
E W Wuis et al.
Journal of chromatography, 526(2), 575-580 (1990-04-06)
Furan derivatives. XXX. preparation of substituted 5-phenyl-2-furfuryl alcohols.
Frimm RAK and Kovac J.
Chemical Papers, 26, 551-556 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service