Skip to Content
Merck
All Photos(1)

Documents

428264

Sigma-Aldrich

N-(4-Methoxyphenyl)acetamide

97%

Synonym(s):

p-Acetanisidide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OCH3
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

97%

mp

128-130 °C (lit.)

SMILES string

COc1ccc(NC(C)=O)cc1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eyal Shteyer et al.
Journal of pediatric gastroenterology and nutrition, 56(1), 60-65 (2012-06-15)
Distinguishing biliary atresia (BA) from other causes of neonatal cholestasis (NC) is challenging. Continuous BreathID C-methacetin breath test (MBT) is a novel method that determines liver function. Methacetin is metabolized uniquely by the liver and CO2 is measured passively, through
A Kochel-Jankowska et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 64(1), 27-33 (2013-04-10)
This prospective study intended to ascertain if cytochrome P450 dependent liver function is affected in early and late histological stages of primary biliary cirrhosis (PBC). The study included 32 female PBC patients (mean age 55.4 years, range 33-70) and 16
Krista J M Stibbe et al.
Scandinavian journal of gastroenterology, 46(7-8), 962-972 (2011-06-01)
Chronic viral hepatitis B and C cause liver fibrosis, leading to cirrhosis. Fibrosis assessment is essential to establish prognosis and treatment indication. We compared seven non-invasive tests, separately and in combination, in chronic hepatitis patients to detect early stages of
J F Lock et al.
European journal of medical research, 14, 547-550 (2010-02-13)
Non-invasive breath tests can serve as valuable diagnostic tools in medicine as they can determine particular enzymatic and metabolic functions in vivo. However, methodological pitfalls have limited the actual clinical application of those tests till today. A major challenge of
Sven Dänicke et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(1), 152-158 (2009-10-03)
A 35-day piglet experiment starting from weaning (21 days of age, 7.7+/-1.1 kg), was performed to examine the effects of feeding a control diet (CTR) and of a Fusarium toxin-contaminated diet (FUS) in the absence (-) or presence (+) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service