Skip to Content
Merck
All Photos(3)

Key Documents

427845

Sigma-Aldrich

Polypropylene-graft-maleic anhydride

viscosity 425 cPs

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$78.90
1 KG
$218.00

$78.90


Estimated to ship onMarch 24, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$78.90
1 KG
$218.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$78.90


Estimated to ship onMarch 24, 2025


Request a Bulk Order

Quality Level

form

beads

composition

maleic anhydride, 8-10 wt. %

viscosity

4.0 poise(190 °C, Brookfield Thermosel)(lit.)

mp

156 °C

acid number

40.0‑55.0(mg-KOH)

transition temp

softening point 157 °C (ring and ball)

density

0.934 g/mL at 25 °C

SMILES string

O1C(=O)C=CC1=O.CC=C

InChI

1S/C4H2O3.C3H6/c5-3-1-2-4(6)7-3;1-3-2/h1-2H;3H,1H2,2H3

InChI key

DBVUAFDZHKSZJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Imparts slip resistance to polishes, coupling additive for filled polypropylene and polymer alloys, pigment dispersant and processing aid for plastics, such as ABS.

Features and Benefits

High melting, hard, emulsifiable wax. Grafted with maleic anhydride to give hydrophilic characteristics.

Physical form

Crystalline, isotactic, modified homopolymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Buongiorno qualche tempo fa acquistai 1Kg di Polypropylene-graft-maleic anhydride e pertanto volevo sapere se tale materia prima fosse idonea secondo quanto previsto dal Regolamento (EU) 10/2011 per il contatto diretto con alimenti.

    1 answer
    1. Unless otherwise stated, this product is intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service