Skip to Content
Merck
All Photos(2)

Key Documents

427470

Sigma-Aldrich

(S)-(−)-N-Benzyl-α-methylbenzylamine

99%

Synonym(s):

(S)-(−)-N-(1-Phenylethyl)benzylamine, (S)-(−)-N-Benzyl-α-phenylethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$116.00
50 ML
$428.00

$116.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$116.00
50 ML
$428.00

About This Item

Linear Formula:
C6H5CH(CH3)NHCH2C6H5
CAS Number:
Molecular Weight:
211.30
Beilstein:
3650264
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$116.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

optical activity

[α]19/D −40°, neat

optical purity

ee: ≥97% (HPLC)

refractive index

n20/D 1.563 (lit.)

bp

171 °C/15 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

C[C@H](NCc1ccccc1)c2ccccc2

InChI

1S/C15H17N/c1-13(15-10-6-3-7-11-15)16-12-14-8-4-2-5-9-14/h2-11,13,16H,12H2,1H3/t13-/m0/s1

InChI key

ZYZHMSJNPCYUTB-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(−)-N-Benzyl-α-methylbenzylamine can be used:
  • In one of the key synthetic steps for the preparation of a cardioprotective drug named CP-060S.[1]
  • To prepare (1S,2S,3S,5R)-tert-butyl 3-[benzyl((S)-1-phenylethyl)amino]-6,6-dimethylbicyclo[3.1.1]heptane-2-carboxylate, an intermediate used to synthesize pinane-based β- and γ-amino acids.[2]
  • To prepare urea derivatives as potent antimicrobial agents.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of pinane-based beta-and gamma-amino acids via conjugate addition of lithium amides and nitromethane
Szakonyi Z, et al.
Tetrahedron Asymmetry, 21(20), 2498-2504 (2010)
New urea derivatives as potential antimicrobial agents: Synthesis, biological evaluation, and molecular docking studies
Patil M, et al.
Antibiotics, 8(4), 178-178 (2019)
Practical Synthesis of Novel Cardioprotective Drug, CP-060 S
Kato T, et al.
Organic Process Research & Development, 5(2), 122-126 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service