Skip to Content
Merck
All Photos(1)

Key Documents

427365

Sigma-Aldrich

2-Biphenylyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4NCO
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.606 (lit.)

bp

92-94 °C/1 mmHg (lit.)

density

1.138 g/mL at 25 °C (lit.)

SMILES string

O=C=Nc1ccccc1-c2ccccc2

InChI

1S/C13H9NO/c15-10-14-13-9-5-4-8-12(13)11-6-2-1-3-7-11/h1-9H

InChI key

IHHUGFJSEJSCGE-UHFFFAOYSA-N

General description

2-Biphenylyl isocyanate is an aryl isocyanate.[1] It has been synthesized from 2-aminobiphenyl and phosgene. The density, viscosity, refractive index and dielectric constant of benzene solution of 2-biphenylyl isocyanate have been measured.[2]

Application

2-Biphenylyl isocyanate may be used in the synthesis of 2-bromophenanthridone.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave Absorption and Molecular Structure in Liquids. XXIII. The Dielectric Relaxation Times and Dipole Moments of Several Aryl Isocyanates and Related Compounds1.
Jolliffe BR and Smyth CP.
Journal of the American Chemical Society, 80(5), 1064-1066 (1958)
2-Bromophenanthridone.
Mosby WL.
Journal of the American Chemical Society, 76(3), 936-937 (1954)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service