Skip to Content
Merck
All Photos(1)

Key Documents

426245

Sigma-Aldrich

Ethylene-d4-diamine

98 atom % D, 98% (CP)

Synonym(s):

Ethylenediamine-1,1,2,2-d4

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$788.00
1 G
$1,990.00

$788.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$788.00
1 G
$1,990.00

About This Item

Linear Formula:
H2NCD2CD2NH2
CAS Number:
Molecular Weight:
64.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$788.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

refractive index

n20/D 1.455 (lit.)

bp

118 °C (lit.)

mp

9 °C (lit.)

density

1.022 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]C([2H])(N)C([2H])([2H])N

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2/i1D2,2D2

InChI key

PIICEJLVQHRZGT-LNLMKGTHSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service