Skip to Content
Merck
All Photos(2)

Documents

423734

Sigma-Aldrich

6-Methyl-1-indanone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

70 °C/0.4 mmHg (lit.)

mp

60-62 °C (lit.)

SMILES string

Cc1ccc2CCC(=O)c2c1

InChI

1S/C10H10O/c1-7-2-3-8-4-5-10(11)9(8)6-7/h2-3,6H,4-5H2,1H3

InChI key

DBOXRDYLMJMQBB-UHFFFAOYSA-N

General description

6-Methyl-1-indanone is a substituted indanone. It has been synthesized in high quantum yields by the photolysis of α-chloro-2′,5′-dimethylacetophenone. It is formed as one of the photoproduct during the irradiation of 2,5-dimethylphenacyl (DMP) esters. It is reported to be one of the semivolatile component of lamina cigarette smoke.

Application

6-Methyl-1-indanone was used as a starting material for the synthesis of branched alkyl indanes (BINs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 5-Dimethylphenacyl as a new photoreleasable protecting group for carboxylic acids.
Klan P, et al.
Organic Letters, 2(11), 1569-1571 (2000)
Comparisons of Smoke Components in the Semivolatile Phase from Lamina and Midrib Cigarettes of Flue-cured Tobacco Leaves.
Ishiguro S and Sugawara S.
Agricultural and Biological Chemistry, 42(8), 1527-1531 (1978)
A laser flash photolysis study of the mechanism of the photocyclization of. alpha.-chloro-ortho-methylacetophenones.
Netto-Ferreira JC and Scaiano JC.
Journal of the American Chemical Society, 113(15), 5800-5803 (1991)
Rapid Photochemical Synthesis of 6-Methyl-1-Indanone.
Wang S, et al.
Advanced Materials Research, 634-638, 416-419 (2013)
Andrew M Booth et al.
Environmental science & technology, 42(21), 8122-8126 (2008-11-27)
Previously, comprehensive two-dimensional gas chromatography-time of flight-mass-spectrometry (GCxGC-ToF-MS) revealed that the unresolved complex mixtures (UCMs) of contaminant hydrocarbons accumulated by health-affected mussels Mytilus edulis (up to 125 microg g dry weight(-1)) collected from around U.K. coasts, included many isomeric branched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service